Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Glass bottle
Catalog number FSA174615000
Price (MYR)
1,080.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,080.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriethoxyvinylsilane
Name Note97%
CAS78-08-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.