2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
Thermo Scientific Chemicals
2,5-Dimethylpyrazine, 99%
CAS: 123-32-0 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA174520050
Price (MYR)
401.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
401.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 5-Dimethylpyrazine
Name Note99%
CAS123-32-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning