Stearic acid, 97%
Stearic acid, 97%
Stearic acid, 97%
Thermo Scientific Chemicals
Stearic acid, 97%
CAS: 57-11-4 | C18H36O2 | 284.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Fibre drum
Catalog number FSA174490100
Price (MYR)
1,447.00
EA
Add to cart
Quantity:
10 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (MYR)
1,447.00
EA
Add to cart
Specifications
Chemical Name or MaterialStearic acid
CAS57-11-4
Melting Point67.0°C to 69.0°C
ColorWhite
Density0.8400g/mL