Check Order Status
Methyl 4-methoxysalicylate, 98%, Thermo Scientific™
Element Search
Structure Search
Methyl 4-methoxysalicylate, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
10 g
Packaging:
Glass bottle
Catalog number
FSA174270100
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Methyl 4-methoxysalicylate
CAS
5446-02-6
Melting Point
49°C to 53°C
Color
White to Yellow
EINECS Number
226-657-5