Tetradecane, 99%
Tetradecane, 99%
Tetradecane, 99%
Thermo Scientific Chemicals
Tetradecane, 99%
CAS: 629-59-4 | C14H30 | 198.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA174135000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetradecane
CAS629-59-4
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Use personal protective equipment as required.
Melting Point5.8°C