Tetraethylene glycol dimethyl ether, 99%
Tetraethylene glycol dimethyl ether, 99%
Tetraethylene glycol dimethyl ether, 99%
Tetraethylene glycol dimethyl ether, 99%
Thermo Scientific Chemicals
Tetraethylene glycol dimethyl ether, 99%
CAS: 143-24-8 | C10H22O5 | 222.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA174112500
Price (MYR)
357.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
357.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetraethylene glycol dimethyl ether
Acidity0.015% max.
CAS143-24-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.