L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
L(+)-Rhamnose monohydrate, 99%
Thermo Scientific Chemicals
L(+)-Rhamnose monohydrate, 99%
CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA174085000
Price (MYR)
6,537.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,537.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Rhamnose monohydrate
CAS10030-85-0
Melting Point91.0°C to 93.0°C
ColorWhite
Ignition Residue0.2% max.