Specifications
Chemical Name or MaterialQuercetin hydrate
CAS849061-97-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
View more