n-Amylamine, 99%
n-Amylamine, 99%
n-Amylamine, 99%
Thermo Scientific Chemicals
n-Amylamine, 99%
CAS: 110-58-7 | C5H13N | 87.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA173942500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Amylamine
CAS110-58-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0