o-Anisic acid, 99%
o-Anisic acid, 99%
o-Anisic acid, 99%
Thermo Scientific Chemicals
o-Anisic acid, 99%
CAS: 579-75-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA173751000
Price (MYR)
320.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
320.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Anisic acid
Name Note99%
CAS579-75-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning