4,4'-Biphenol, 97%
4,4'-Biphenol, 97%
4,4'-Biphenol, 97%
Thermo Scientific Chemicals
4,4'-Biphenol, 97%
CAS: 92-88-6 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
View all
Plastic bottle
Catalog number FSA173671000
Price (MYR)
942.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
942.00
EA
Add to cart
Specifications
Chemical Name or Material4, 4'-Biphenol
CAS92-88-6
CAS Min %96.0
CAS Max %100.0
Health Hazard 2GHS H Statement: