Celite™, for analysis, high purity analytical grade
Celite™, for analysis, high purity analytical grade
Celite™, for analysis, high purity analytical grade
Celite™, for analysis, high purity analytical grade
Thermo Scientific Chemicals
Celite™, for analysis, high purity analytical grade
CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass Bottle
Catalog number FSA173460250
Price (MYR)
1,118.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,118.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCelite™
Density<0.25 g/mL (centrifuged wet density)
Physical FormPowder
ColorGray/Pink to White
Quantity25 g
View more
Filter aid
RUO – Research Use Only