Adipoyl chloride, 98%
Adipoyl chloride, 98%
Adipoyl chloride, 98%
Adipoyl chloride, 98%
Thermo Scientific Chemicals
Adipoyl chloride, 98%
Adipoyl chloride, 98%, C6H8Cl2O2, CAS Number-111-50-2, adipic acid chloride, adipic acid dichloride, adipic dichloride, adipoylchloride, adipoyl chloride, adipoyl dichloride, adipyl chloride, butane-1,4-dicarbonyl chloride, hexanedioyl chloride, pubchem13242, 500mL, 02, 653 | CAS: 111-50-2 | C6H8Cl2O2 | 183.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA173185000
Price (MYR)
3,840.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,840.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAdipoyl chloride
Name Note98%
CAS111-50-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger