Abietic acid, 85%
Abietic acid, 85%
Abietic acid, 85%
Thermo Scientific Chemicals
Abietic acid, 85%
CAS: 514-10-3 | C20H30O2 | 302.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA173130250
Price (MYR)
522.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
522.00
EA
Add to cart
Specifications
Chemical Name or MaterialAbietic acid
Name Note85%
CAS514-10-3
CAS Min %80.0
CAS Max %100.0