Tricine, 98%, for biochemistry
Tricine, 98%, for biochemistry
Tricine, 98%, for biochemistry
Tricine, 98%, for biochemistry
Thermo Scientific Chemicals
Tricine, 98%, for biochemistry
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA172641000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance0.05 max. at 250nm
Chemical Name or MaterialTricine
Absorbance Conditions(5% aq. soln.) (1 cm cell)
Name Note98%
CAS5704-04-1