CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
CAPS, 99%, for biochemistry
Thermo Scientific Chemicals
CAPS, 99%, for biochemistry
CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA172621000
Price (MYR)
501.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
BOT
Add to cart
Specifications
Absorbance(0.1 M in H2O) at 0nm
Chemical Name or MaterialCAPS
Absorbance Conditions(0.1M in H2O)
Name Note99%
CAS1135-40-6
View more
Biological buffer
RUO – Research Use Only