EPPS, 99%, for biochemistry
EPPS, 99%, for biochemistry
EPPS, 99%, for biochemistry
EPPS, 99%, for biochemistry
Thermo Scientific Chemicals
EPPS, 99%, for biochemistry
CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA172601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance(0.1 M aq. soln.) (1 cm cell)
Chemical Name or MaterialEPPS
Absorbance Conditions(0.1M aq. soln.) (1 cm cell)
CAS16052-06-5
ColorWhite
View more
Biological buffer
RUO – Research Use Only