MES hydrate, 99+%, for biochemistry
MES hydrate, 99+%, for biochemistry
MES hydrate, 99+%, for biochemistry
MES hydrate, 99+%, for biochemistry
MES hydrate, 99+%, for biochemistry
MES hydrate, 99+%, for biochemistry
Thermo Scientific Chemicals
MES hydrate, 99+%, for biochemistry
CAS: 1266615-59-1 | C6H13NO4S | 195.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA172590250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance(0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Chemical Name or MaterialMES hydrate
Absorbance Conditions(0.1M aq. soln.) (1 cm cell)
Name Note0.99
CAS1266615-59-1