BES, 99+%, for biochemistry
BES, 99+%, for biochemistry
BES, 99+%, for biochemistry
BES, 99+%, for biochemistry
Thermo Scientific Chemicals
BES, 99+%, for biochemistry
CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA172585000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance0.095 max. (1 M in H2O) at 260nm
Chemical Name or MaterialBES
CAS10191-18-1
Melting Point152.0°C to 154.0°C
CAS Min %99.0
View more
Biological buffer
RUO – Research Use Only