HEPES, 99%, for biochemistry
HEPES, 99%, for biochemistry
HEPES, 99%, for biochemistry
HEPES, 99%, for biochemistry
Thermo Scientific Chemicals
HEPES, 99%, for biochemistry
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA172572500
Price (MYR)
1,242.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,242.00
BOT
Add to cart
Specifications
Absorbance0.08 max. at 260nm, 0.05 max. at 280nm, 0.08 max. at 250nm, (5% in water) 1 cm cell, (5% in water) 1 cm cell at 0nm
Chemical Name or MaterialHEPES
Absorbance Conditions(5% aq. soln.) (1cm cell)
CAS7365-45-9
Melting Point234.0°C to 238.0°C
View more
Biological buffer
RUO – Research Use Only