2-Picolyl chloride hydrochloride, 98%
2-Picolyl chloride hydrochloride, 98%
2-Picolyl chloride hydrochloride, 98%
2-Picolyl chloride hydrochloride, 98%
Thermo Scientific Chemicals
2-Picolyl chloride hydrochloride, 98%
CAS: 6959-47-3 | C6H6ClN·HCl | 164.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA172120250
Price (MYR)
530.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
530.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Picolyl chloride hydrochloride
Name Note98%
CAS6959-47-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger