D-Alanine, 99+%
D-Alanine, 99+%
D-Alanine, 99+%
Thermo Scientific Chemicals
D-Alanine, 99+%
CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA172080050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Alanine
Melting Point278.0°C to 282.0°C
CAS338-69-2
Arsenic (As)2ppm max. (As2O3)