D-Phenylalanine, 99+%
D-Phenylalanine, 99+%
D-Phenylalanine, 99+%
D-Phenylalanine, 99+%
Thermo Scientific Chemicals
D-Phenylalanine, 99+%
CAS: 673-06-3 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass Bottle
Catalog number FSA172041000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Phenylalanine
CAS673-06-3
Arsenic (As)3ppm max.
ColorWhite
Heavy Metals (as Pb)20ppm max.