2-Amino-6-methoxybenzothiazole, 98%
2-Amino-6-methoxybenzothiazole, 98%
2-Amino-6-methoxybenzothiazole, 98%
2-Amino-6-methoxybenzothiazole, 98%
Thermo Scientific Chemicals
2-Amino-6-methoxybenzothiazole, 98%
CAS: 1747-60-0 | C8H8N2OS | 180.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA17202050050 gGlass bottle
Catalog number FSA172020500
Price (MYR)
490.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-6-methoxybenzothiazole
Name Note98%
CAS1747-60-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger