4-Fluorobenzylamine, 97%
4-Fluorobenzylamine, 97%
4-Fluorobenzylamine, 97%
4-Fluorobenzylamine, 97%
Thermo Scientific Chemicals
4-Fluorobenzylamine, 97%
CAS: 140-75-0 | C7H8FN | 125.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA171950250
Price (MYR)
517.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
517.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Fluorobenzylamine
CAS140-75-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat