4'-Bromoacetanilide, 98%
4'-Bromoacetanilide, 98%
4'-Bromoacetanilide, 98%
4'-Bromoacetanilide, 98%
Thermo Scientific Chemicals
4'-Bromoacetanilide, 98%
CAS: 103-88-8 | C8H8BrNO | 214.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA171570250
Price (MYR)
329.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
329.00
EA
Add to cart
Specifications
Chemical Name or Material4′-Bromoacetanilide
CAS103-88-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0