Activated charcoal, NORIT™ SA 2, decolorizing
Activated charcoal, NORIT™ SA 2, decolorizing
Activated charcoal, NORIT™ SA 2, decolorizing
Activated charcoal, NORIT™ SA 2, decolorizing
Thermo Scientific Chemicals
Activated charcoal, NORIT™ SA 2, decolorizing
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
Plastic drum
Catalog number FSA171530050
Price (MYR)
1,549.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,549.00
EA
Add to cart
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* SA 2
CAS7440-44-0
Melting Point3652.0°C
ColorBlack
EINECS Number231-153-3
View more
Filter aid
RUO – Research Use Only