1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
Thermo Scientific Chemicals
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
CAS: 25952-53-8 | C8H18ClN3 | 191.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 g
10 g
50 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA171445000
Price (MYR)
15,881.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
15,881.00
EA
Add to cart
Specifications
Chemical Name or Material1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride
CAS25952-53-8
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only