meso-Tetraphenylporphine, 97%
meso-Tetraphenylporphine, 97%
meso-Tetraphenylporphine, 97%
meso-Tetraphenylporphine, 97%
Thermo Scientific Chemicals
meso-Tetraphenylporphine, 97%
CAS: 917-23-7 | C44H30N4 | 614.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA171210250
Price (MYR)
4,658.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,658.00
EA
Add to cart
Specifications
Chemical Name or Materialmeso-Tetraphenylporphine
CAS917-23-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W