Succinonitrile, 99+%
Succinonitrile, 99+%
Succinonitrile, 99+%
Succinonitrile, 99+%
Thermo Scientific Chemicals
Succinonitrile, 99+%
CAS: 110-61-2 | C4H4N2 | 80.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA171185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSuccinonitrile
Name Note99+%
CAS110-61-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement