Iodoacetic acid, sodium salt, 99%
Iodoacetic acid, sodium salt, 99%
Iodoacetic acid, sodium salt, 99%
Iodoacetic acid, sodium salt, 99%
Thermo Scientific Chemicals
Iodoacetic acid, sodium salt, 99%
CAS: 305-53-3 | C2H2INaO2 | 207.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA170971000
Price (MYR)
2,524.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,524.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodoacetic acid, sodium salt
CAS305-53-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.