N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%
Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%
CAS: 100-22-1 | C10H16N2 | 164.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA170790250
Price (MYR)
1,815.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,815.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N,N',N'-Tetramethyl-p-phenylenediamine
CAS100-22-1
Health Hazard 1GHS Signal Word: Warning
Melting Point48.0°C to 52.0°C
ColorBeige-Brown to Brown