2-Bromo-4'-methylacetophenone, 97%
2-Bromo-4'-methylacetophenone, 97%
2-Bromo-4'-methylacetophenone, 97%
2-Bromo-4'-methylacetophenone, 97%
Thermo Scientific Chemicals
2-Bromo-4'-methylacetophenone, 97%
CAS: 619-41-0 | C9H9BrO | 213.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA170390250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-4'-methylacetophenone
CAS619-41-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0