Diethyl pyrocarbonate, 97%, pure
Diethyl pyrocarbonate, 97%, pure
Diethyl pyrocarbonate, 97%, pure
Diethyl pyrocarbonate, 97%, pure
Thermo Scientific Chemicals
Diethyl pyrocarbonate, 97%, pure
CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA170255000
Price (MYR)
5,933.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,933.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethyl pyrocarbonate
CAS1609-47-8
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Flammable liquids (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Health Hazard 2Causes serious eye irritation, Harmful if swallowed, Combustible liquid, May cause respiratory irritation, Causes skin irritation
Density1.1175g/mL