Methyl p-toluenesulfonate, 97%
Methyl p-toluenesulfonate, 97%
Methyl p-toluenesulfonate, 97%
Methyl p-toluenesulfonate, 97%
Thermo Scientific Chemicals
Methyl p-toluenesulfonate, 97%
CAS: 80-48-8 | C8H10O3S | 186.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA170101000
Price (MYR)
273.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
273.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl p-toluenesulfonate
Name Note97%
Acidity0.2% max.
CAS80-48-8
Health Hazard 1GHS Signal Word: Danger