Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA170050025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriethanolamine hydrochloride
Name Notep.a.
CAS637-39-8
Health Hazard 1HYGROSCOPIC