4'-Chloroacetanilide, 97%
4'-Chloroacetanilide, 97%
4'-Chloroacetanilide, 97%
4'-Chloroacetanilide, 97%
Thermo Scientific Chemicals
4'-Chloroacetanilide, 97%
CAS: 539-03-7 | C8H8ClNO | 169.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA169950500
Price (MYR)
688.00
EA
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
688.00
EA
Add to cart
Specifications
Chemical Name or Material4′-Chloroacetanilide
CAS539-03-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0