Phenoxyacetic acid, 98+%
Phenoxyacetic acid, 98+%
Phenoxyacetic acid, 98+%
Phenoxyacetic acid, 98+%
Thermo Scientific Chemicals
Phenoxyacetic acid, 98+%
CAS: 122-59-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA169875000
Price (MYR)
476.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
476.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenoxyacetic acid
CAS122-59-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth