8-Pentadecanone, 98%
8-Pentadecanone, 98%
8-Pentadecanone, 98%
8-Pentadecanone, 98%
Thermo Scientific Chemicals
8-Pentadecanone, 98%
CAS: 818-23-5 | C15H30O | 226.40 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA16979025025 gGlass bottle
Catalog number FSA169790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material8-Pentadecanone
Name Note98%
CAS818-23-5
CAS Min %97.5
CAS Max %100.0