4-(Benzyloxy)phenol, 98%
4-(Benzyloxy)phenol, 98%
4-(Benzyloxy)phenol, 98%
4-(Benzyloxy)phenol, 98%
Thermo Scientific Chemicals
4-(Benzyloxy)phenol, 98%
CAS: 103-16-2 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA169780500
Price (MYR)
420.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
420.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Benzyloxy)phenol
Name Note98%
CAS103-16-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning