Methyl (triphenylphosphoranylidene)acetate, 98%
Methyl (triphenylphosphoranylidene)acetate, 98%
Methyl (triphenylphosphoranylidene)acetate, 98%
Methyl (triphenylphosphoranylidene)acetate, 98%
Thermo Scientific Chemicals
Methyl (triphenylphosphoranylidene)acetate, 98%
CAS: 2605-67-6 | C21H19O2P | 334.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA169560250
Price (MYR)
545.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
545.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl (triphenylphosphoranylidene)acetate
CAS2605-67-6
Health Hazard 1GHS Signal Word: Danger
Melting Point164.0°C to 169.0°C
ColorWhite
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only