4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
Thermo Scientific Chemicals
4-tert-Butylbenzoyl chloride, 98%
CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA16910025025 gGlass bottle
Catalog number FSA169100250
Price (MYR)
559.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
559.00
BOT
Add to cart
Specifications
Chemical Name or Material4-tert-Butylbenzoyl chloride
Name Note98%
CAS1710-98-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger