2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
Thermo Scientific Chemicals
2-(Ethylamino)ethanol, 98%
CAS: 110-73-6 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA168732500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(Ethylamino)ethanol
Name Note98%
CAS110-73-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger