Hydantoin, 99%
Hydantoin, 99%
Hydantoin, 99%
Thermo Scientific Chemicals
Hydantoin, 99%
CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA168701000
Price (MYR)
333.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
333.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydantoin
Name Note99%
CAS461-72-3
Melting Point220°C to 223°C
ColorWhite to Yellow