4-Fluoro-3-nitroaniline, 99%
4-Fluoro-3-nitroaniline, 99%
4-Fluoro-3-nitroaniline, 99%
4-Fluoro-3-nitroaniline, 99%
Thermo Scientific Chemicals
4-Fluoro-3-nitroaniline, 99%
CAS: 364-76-1 | C6H5FN2O2 | 156.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA168400250
Price (MYR)
278.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
278.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Fluoro-3-nitroaniline
Name Note99%
CAS364-76-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning