2,4,6-Tri(2-pyridyl)-s-triazine, 99%
2,4,6-Tri(2-pyridyl)-s-triazine, 99%
2,4,6-Tri(2-pyridyl)-s-triazine, 99%
2,4,6-Tri(2-pyridyl)-s-triazine, 99%
Thermo Scientific Chemicals
2,4,6-Tri(2-pyridyl)-s-triazine, 99%
CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA168071000
Price (MYR)
10,153.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
10,153.00
EA
Add to cart
Specifications
Chemical Name or Material2, 4, 6-Tri(2-pyridyl)-s-triazine
CAS3682-35-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0