Tetrachloroethylene, 99+%, spectrophotometric grade
Tetrachloroethylene, 99+%, spectrophotometric grade
Tetrachloroethylene, 99+%, spectrophotometric grade
Tetrachloroethylene, 99+%, spectrophotometric grade
Thermo Scientific Chemicals
Tetrachloroethylene, 99+%, spectrophotometric grade
CAS: 127-18-4 | C2Cl4 | 165.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA167890010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrachloroethylene
CAS127-18-4
Absorbance(1cm cell vs water), (1 cm cell vs water) at 0nm, 0.03 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 305nm, 0.3 max. at 295nm, 1 max. at 290nm
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Causes serious eye irritati