Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass Bottle
Catalog number FSA167851000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
Name Note99.9%
Absorbance0.01 max. at 360nm, 0.05 max. at 320nm, 0.1 max. at 300nm, 0.2 max. at 290nm, 0.3 max. at 280nm, 0.5 max. at 270nm
CAS67-68-5
View more
For UV/VIS spectroscopy
RUO – Research Use Only