Methanol, 99.9%, for spectroscopy
Methanol, 99.9%, for spectroscopy
Methanol, 99.9%, for spectroscopy
Methanol, 99.9%, for spectroscopy
Thermo Scientific Chemicals
Methanol, 99.9%, for spectroscopy
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA167835000
Price (MYR)
333.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
333.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
Absorbance0.01 max. at 260nm, 0.05 max. at 240nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 0.6 max. at 210nm, 1 max. at 205nm
CAS67-56-1
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only