n-Heptane, for HPLC, approx. 95% n-heptane
n-Heptane, for HPLC, approx. 95% n-heptane
n-Heptane, for HPLC, approx. 95% n-heptane
n-Heptane, for HPLC, approx. 95% n-heptane
Thermo Scientific Chemicals
n-Heptane, for HPLC, approx. 95% n-heptane
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA167820010
Price (MYR)
415.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
415.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.01 max. at 245nm, 0.046 max. at 227nm, 0.10 max. at 220nm, 0.301 max. at 210nm, 0.7 max. at 200nm
CAS142-82-5
Health Hazard 1Danger
View more
For HPLC
RUO – Research Use Only